Product Details
- SNP ID
-
rs145497205
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:7763294 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTGACTCCAAGGAACCAAGGGTG[A/C]AGCAGCTGGGCCTCCTGGGTAAGGC
- Phenotype
-
MIM: 605872
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLEC4M
PubMed Links
Gene Details
- Gene
- CLEC4M
- Gene Name
- C-type lectin domain family 4 member M
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001144904.1 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
NP_001138376.1 |
NM_001144905.1 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
NP_001138377.1 |
NM_001144906.1 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
NP_001138378.1 |
NM_001144907.1 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
NP_001138379.1 |
NM_001144908.1 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
NP_001138380.1 |
NM_001144909.1 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
NP_001138381.1 |
NM_001144910.1 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
NP_001138382.1 |
NM_001144911.1 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
NP_001138383.1 |
NM_014257.4 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
NP_055072.3 |
XM_006722611.2 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
XP_006722674.1 |
XM_006722612.3 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
XP_006722675.1 |
XM_006722613.3 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
XP_006722676.1 |
XM_006722614.3 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
XP_006722677.1 |
XM_006722615.2 |
146 |
Missense Mutation |
AAG,CAG |
K10Q |
XP_006722678.1 |
View Full Product Details