Product Details

SNP ID
rs145501224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17421562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTATAGTAAATATTTTAGGCTTTA[C/T]AGGGCAGATGATGTGTTTTGCATAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BISPR PubMed Links

Gene Details

Gene
BISPR
Gene Name
BST2 interferon stimulated positive regulator (non-protein coding)
There are no transcripts associated with this gene.

Gene
MVB12A
Gene Name
multivesicular body subunit 12A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304547.1 Intron NP_001291476.1
NM_138401.3 Intron NP_612410.1

View Full Product Details