Product Details
- SNP ID
-
rs145590794
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:18060064 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGATGCCTCTTCCTGGAAGTCCA[A/C]TGGGTTGATCCACTGCCAAGTCTGC
- Phenotype
-
MIM: 601604
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IL12RB1
PubMed Links
Gene Details
- Gene
- IL12RB1
- Gene Name
- interleukin 12 receptor subunit beta 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001290023.1 |
2228 |
Missense Mutation |
GTG,TTG |
V605L |
NP_001276952.1 |
NM_001290024.1 |
2228 |
Missense Mutation |
GTG,TTG |
V645L |
NP_001276953.1 |
NM_005535.2 |
2228 |
Missense Mutation |
GTG,TTG |
V605L |
NP_005526.1 |
NM_153701.2 |
2228 |
Intron |
|
|
NP_714912.1 |
XM_006722741.3 |
2228 |
Missense Mutation |
GTG,TTG |
V645L |
XP_006722804.2 |
XM_011527966.2 |
2228 |
Missense Mutation |
GTG,TTG |
V656L |
XP_011526268.1 |
XM_011527967.2 |
2228 |
Missense Mutation |
GTG,TTG |
V652L |
XP_011526269.1 |
XM_011527968.2 |
2228 |
Missense Mutation |
GTG,TTG |
V649L |
XP_011526270.1 |
XM_011527969.2 |
2228 |
Missense Mutation |
GTG,TTG |
V645L |
XP_011526271.1 |
XM_011527970.2 |
2228 |
Missense Mutation |
GTG,TTG |
V656L |
XP_011526272.1 |
XM_011527971.2 |
2228 |
Missense Mutation |
GTG,TTG |
V656L |
XP_011526273.1 |
XM_011527972.2 |
2228 |
Missense Mutation |
GTG,TTG |
V656L |
XP_011526274.1 |
XM_011527973.2 |
2228 |
Missense Mutation |
GTG,TTG |
V616L |
XP_011526275.1 |
XM_011527974.2 |
2228 |
Missense Mutation |
GTG,TTG |
V612L |
XP_011526276.1 |
XM_011527975.2 |
2228 |
Missense Mutation |
GTG,TTG |
V645L |
XP_011526277.1 |
XM_011527976.2 |
2228 |
Intron |
|
|
XP_011526278.1 |
XM_011527977.2 |
2228 |
Intron |
|
|
XP_011526279.1 |
XM_017026762.1 |
2228 |
Missense Mutation |
GTG,TTG |
V411L |
XP_016882251.1 |
View Full Product Details