Product Details

SNP ID
rs145686572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50625919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTGAGAAGCCAGTGAGGTCCAT[C/T]GCTTTGAGGTTAGAGGCTTTGATGA
Phenotype
MIM: 600327
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SYT3 PubMed Links

Gene Details

Gene
SYT3
Gene Name
synaptotagmin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160328.1 1579 Silent Mutation GCA,GCG A460A NP_001153800.1
NM_001160329.1 1579 Silent Mutation GCA,GCG A460A NP_001153801.1
NM_032298.2 1579 Silent Mutation GCA,GCG A460A NP_115674.1
XM_011527390.2 1579 Silent Mutation GCA,GCG A460A XP_011525692.1
XM_011527391.2 1579 Silent Mutation GCA,GCG A460A XP_011525693.1

View Full Product Details