Product Details

SNP ID
rs145722764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45517949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGTGGTCATCAACTGTGCCAT[A/C]GTCCGGGGTGTCAAGTATAACCAGG
Phenotype
MIM: 606580 MIM: 601703
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OPA3 PubMed Links

Gene Details

Gene
OPA3
Gene Name
optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia)
There are no transcripts associated with this gene.

Gene
VASP
Gene Name
vasodilator-stimulated phosphoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003370.3 528 Silent Mutation ATA,ATC I66I NP_003361.1
XM_005259199.1 528 Silent Mutation ATA,ATC I66I XP_005259256.1
XM_005259200.1 528 Silent Mutation ATA,ATC I66I XP_005259257.1
XM_017027200.1 528 Silent Mutation ATA,ATC I66I XP_016882689.1

View Full Product Details