Product Details

SNP ID
rs145762717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49665841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGCCGATGGGACGGCCCGCTGG[A/G]CTGTTCCCGCCCCTATGCCCTTTTT
Phenotype
MIM: 610837 MIM: 603734
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCL2L12 PubMed Links

Gene Details

Gene
BCL2L12
Gene Name
BCL2 like 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040668.1 35 Silent Mutation GGA,GGG G6G NP_001035758.1
NM_001282516.1 35 Silent Mutation GGA,GGG G6G NP_001269445.1
NM_001282517.1 35 Silent Mutation GGA,GGG G6G NP_001269446.1
NM_001282519.1 35 Silent Mutation GGA,GGG G6G NP_001269448.1
NM_001282520.1 35 Silent Mutation GGA,GGG G6G NP_001269449.1
NM_001282521.1 35 Silent Mutation GGA,GGG G6G NP_001269450.1
NM_138639.1 35 Silent Mutation GGA,GGG G6G NP_619580.1
XM_017027345.1 35 Silent Mutation GGA,GGG G6G XP_016882834.1
XM_017027346.1 35 Silent Mutation GGA,GGG G6G XP_016882835.1
Gene
IRF3
Gene Name
interferon regulatory factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197122.1 35 UTR 5 NP_001184051.1
NM_001197123.1 35 UTR 5 NP_001184052.1
NM_001197124.1 35 UTR 5 NP_001184053.1
NM_001197125.1 35 UTR 5 NP_001184054.1
NM_001197126.1 35 UTR 5 NP_001184055.1
NM_001197127.1 35 UTR 5 NP_001184056.1
NM_001197128.1 35 UTR 5 NP_001184057.1
NM_001571.5 35 UTR 5 NP_001562.1
XM_006723197.1 35 UTR 5 XP_006723260.1
XM_006723198.1 35 Intron XP_006723261.1
XM_006723200.1 35 Intron XP_006723263.1
XM_006723201.1 35 Intron XP_006723264.1
XM_006723202.2 35 UTR 5 XP_006723265.1
XM_017026766.1 35 UTR 5 XP_016882255.1
XM_017026767.1 35 Intron XP_016882256.1
Gene
SCAF1
Gene Name
SR-related CTD associated factor 1
There are no transcripts associated with this gene.

View Full Product Details