Product Details

SNP ID
rs145809391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5914713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGCAATGGCAGCGGGACGCTGGA[A/T]CTGGAGGAGTTCCTTCGGGCGCTGC
Phenotype
MIM: 114212 MIM: 603327
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CAPS PubMed Links

Gene Details

Gene
CAPS
Gene Name
calcyphosine
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004058.4 922 Missense Mutation GAA,GAT E164D NP_004049.2
NM_080590.3 922 Missense Mutation GAA,GAT E164D NP_542157.2
Gene
RANBP3
Gene Name
RAN binding protein 3
There are no transcripts associated with this gene.

Gene
VMAC
Gene Name
vimentin-type intermediate filament associated coiled-coil protein
There are no transcripts associated with this gene.

View Full Product Details