Product Details
- SNP ID
-
rs145867502
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:10984227 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCGGGCCCTGGCCCTTCCCCTGGA[A/G]CCATGCTGGGCCCTAGCCCGGGTCC
- Phenotype
-
MIM: 603254
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SMARCA4
PubMed Links
Gene Details
- Gene
- SMARCA4
- Gene Name
- SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001128844.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
NP_001122316.1 |
NM_001128845.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
NP_001122317.1 |
NM_001128846.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
NP_001122318.1 |
NM_001128847.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
NP_001122319.1 |
NM_001128848.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
NP_001122320.1 |
NM_001128849.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
NP_001122321.1 |
NM_003072.3 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
NP_003063.2 |
XM_006722845.2 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
XP_006722908.1 |
XM_006722846.2 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
XP_006722909.1 |
XM_011528198.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
XP_011526500.1 |
XM_017027160.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
XP_016882649.1 |
XM_017027161.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
XP_016882650.1 |
XM_017027162.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
XP_016882651.1 |
XM_017027163.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
XP_016882652.1 |
XM_017027164.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
XP_016882653.1 |
XM_017027165.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
XP_016882654.1 |
XM_017027166.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
XP_016882655.1 |
XM_017027167.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
XP_016882656.1 |
XM_017027168.1 |
357 |
Missense Mutation |
ACC,GCC |
T26A |
XP_016882657.1 |
View Full Product Details