Product Details

SNP ID
rs145894974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14942079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTGACACCTTCATCAATAACAT[C/T]GTGATGTATTTTGTGACCATTGTCC
Phenotype
MIM: 600637
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR7C2 PubMed Links

Gene Details

Gene
OR7C2
Gene Name
olfactory receptor family 7 subfamily C member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012377.1 591 Silent Mutation ATC,ATT I197I NP_036509.1
Gene
SLC1A6
Gene Name
solute carrier family 1 member 6
There are no transcripts associated with this gene.

View Full Product Details