Product Details

SNP ID
rs145985086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17825220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCACTGTCACACTGCATGGAAGG[A/G]GCAAAACTGTAGGCTCCAATACTTG
Phenotype
MIM: 146738 MIM: 600173
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INSL3 PubMed Links

Gene Details

Gene
INSL3
Gene Name
insulin like 3
There are no transcripts associated with this gene.

Gene
JAK3
Gene Name
Janus kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000215.3 4998 UTR 3 NP_000206.2
XM_005259896.3 4998 UTR 3 XP_005259953.1
XM_011527990.1 4998 Intron XP_011526292.1
XM_011527991.2 4998 Intron XP_011526293.1

View Full Product Details