Product Details

SNP ID
rs146022713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58084742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGCCCTCCACGCTCTCTGGGGG[A/T]CCGCCCGCCCTAGCCCCCGCCTGGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN18 PubMed Links

Gene Details

Gene
ZSCAN18
Gene Name
zinc finger and SCAN domain containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145542.1 1674 Silent Mutation GGA,GGT G548G NP_001139014.1
NM_001145543.1 1674 Silent Mutation GGA,GGT G492G NP_001139015.1
NM_001145544.1 1674 Silent Mutation GGA,GGT G356G NP_001139016.1
NM_023926.4 1674 Silent Mutation GGA,GGT G492G NP_076415.3
XM_005259174.4 1674 Silent Mutation GGA,GGT G492G XP_005259231.1
XM_006723335.2 1674 Silent Mutation GGA,GGT G492G XP_006723398.1
XM_011527237.2 1674 Silent Mutation GGA,GGT G547G XP_011525539.1
XM_011527238.1 1674 Silent Mutation GGA,GGT G492G XP_011525540.1
XM_011527239.2 1674 Silent Mutation GGA,GGT G492G XP_011525541.1
XM_017027169.1 1674 Silent Mutation GGA,GGT G491G XP_016882658.1
XM_017027170.1 1674 Silent Mutation GGA,GGT G491G XP_016882659.1
XM_017027171.1 1674 Silent Mutation GGA,GGT G491G XP_016882660.1

View Full Product Details