Product Details

SNP ID
rs146067125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47319901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGTTCCAGACATCCTGGCCTTG[G/T]TCATCTTTGCAGTCGTCTTCCTGGT
Phenotype
MIM: 113995
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C5AR1 PubMed Links

Gene Details

Gene
C5AR1
Gene Name
complement component 5a receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001736.3 146 Missense Mutation GTC,TTC V42F NP_001727.1
XM_005259190.4 146 UTR 5 XP_005259247.1

View Full Product Details