Product Details

SNP ID
rs146073962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54906690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAAACCCCCTGAGCGGATTAAC[A/G]AAGTCCAATTCTACATCCCGGACAT
Phenotype
MIM: 604530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCR1 PubMed Links

Gene Details

Gene
NCR1
Gene Name
natural cytotoxicity triggering receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145457.2 372 Missense Mutation AAA,GAA K80E NP_001138929.2
NM_001145458.2 372 Missense Mutation AAA,GAA K80E NP_001138930.2
NM_001242356.2 372 Intron NP_001229285.1
NM_001242357.2 372 Intron NP_001229286.1
NM_004829.6 372 Missense Mutation AAA,GAA K80E NP_004820.2
XM_011527528.2 372 Missense Mutation AAA,GAA K80E XP_011525830.1
XM_011527529.2 372 Missense Mutation AAA,GAA K80E XP_011525831.1
XM_011527530.2 372 Missense Mutation AAA,GAA K80E XP_011525832.1

View Full Product Details