Product Details

SNP ID
rs146123895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54155912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCTGCTTCCCTCCGCAGACGTT[C/G]AGCTCGAAGCTGGTCCAGGGATGGA
Phenotype
MIM: 604910
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CNOT3 PubMed Links

Gene Details

Gene
CNOT3
Gene Name
CCR4-NOT transcription complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014516.3 652 Intron NP_055331.1
XM_005278279.1 652 Intron XP_005278336.1
XM_005278280.2 652 Intron XP_005278337.1
XM_005278281.1 652 Intron XP_005278338.1
XM_005278282.2 652 Intron XP_005278339.1
XM_011526992.1 652 Intron XP_011525294.1
XM_011526993.2 652 Intron XP_011525295.1
Gene
LENG1
Gene Name
leukocyte receptor cluster member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024316.2 652 Missense Mutation CAA,GAA Q202E NP_077292.2
Gene
TMC4
Gene Name
transmembrane channel like 4
There are no transcripts associated with this gene.

View Full Product Details