Product Details

SNP ID
rs146282647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46019619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAGTTCCAGCCACGGCCCTCGT[A/G]TACGAGCCCGTCTTCTCCAATCAGG
Phenotype
MIM: 604963
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC61 PubMed Links

Gene Details

Gene
CCDC61
Gene Name
coiled-coil domain containing 61
There are no transcripts associated with this gene.

Gene
MIR769
Gene Name
microRNA 769
There are no transcripts associated with this gene.

Gene
PGLYRP1
Gene Name
peptidoglycan recognition protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005091.2 593 Missense Mutation CAC,TAC H106Y NP_005082.1

View Full Product Details