Product Details
- SNP ID
-
rs146297522
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:51965073 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCCACAGAAGGCACTTGTGTAGT[C/T]GCCCCGTTAGCAGAGTTTTTCTCCT
- Phenotype
-
MIM: 605422
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF350
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4986771,rs4988336] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF350
- Gene Name
- zinc finger protein 350
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021632.3 |
1767 |
Silent Mutation |
GCA,GCG |
A460A |
NP_067645.3 |
XM_011527196.1 |
1767 |
Silent Mutation |
GCA,GCG |
A424A |
XP_011525498.1 |
XM_017027094.1 |
1767 |
Silent Mutation |
GCA,GCG |
A502A |
XP_016882583.1 |
XM_017027095.1 |
1767 |
Silent Mutation |
GCA,GCG |
A502A |
XP_016882584.1 |
XM_017027096.1 |
1767 |
Silent Mutation |
GCA,GCG |
A502A |
XP_016882585.1 |
XM_017027097.1 |
1767 |
Silent Mutation |
GCA,GCG |
A466A |
XP_016882586.1 |
XM_017027098.1 |
1767 |
Silent Mutation |
GCA,GCG |
A460A |
XP_016882587.1 |
XM_017027099.1 |
1767 |
Silent Mutation |
GCA,GCG |
A460A |
XP_016882588.1 |
XM_017027100.1 |
1767 |
Silent Mutation |
GCA,GCG |
A460A |
XP_016882589.1 |
XM_017027101.1 |
1767 |
Silent Mutation |
GCA,GCG |
A234A |
XP_016882590.1 |
XM_017027102.1 |
1767 |
Silent Mutation |
GCA,GCG |
A234A |
XP_016882591.1 |
XM_017027103.1 |
1767 |
Silent Mutation |
GCA,GCG |
A234A |
XP_016882592.1 |
XM_017027104.1 |
1767 |
Silent Mutation |
GCA,GCG |
A234A |
XP_016882593.1 |
- Gene
- ZNF350-AS1
- Gene Name
- ZNF350 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details