Product Details

SNP ID
rs146311500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49862069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTTCACGTGGACATATCCGGCC[A/G]ACACGAGGTGCTTCTTGAGAAAGGT
Phenotype
MIM: 610221 MIM: 605610 MIM: 610195
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKT1S1 PubMed Links

Gene Details

Gene
AKT1S1
Gene Name
AKT1 substrate 1
There are no transcripts associated with this gene.

Gene
MIR4749
Gene Name
microRNA 4749
There are no transcripts associated with this gene.

Gene
PNKP
Gene Name
polynucleotide kinase 3'-phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007254.3 1273 Missense Mutation NP_009185.2
Gene
PTOV1
Gene Name
prostate tumor overexpressed 1
There are no transcripts associated with this gene.

Gene
PTOV1-AS2
Gene Name
PTOV1 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details