Product Details

SNP ID
rs146338629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5692091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGGAAGGCGATGTCGAAGATCT[C/G]CCGGTAGTGTTCCACGAAGTGCACC
Phenotype
MIM: 605490
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HSD11B1L PubMed Links

Gene Details

Gene
HSD11B1L
Gene Name
hydroxysteroid 11-beta dehydrogenase 1 like
There are no transcripts associated with this gene.

Gene
LONP1
Gene Name
lon peptidase 1, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276479.1 2662 Missense Mutation CAG,GAG Q877E NP_001263408.1
NM_001276480.1 2662 Missense Mutation CAG,GAG Q745E NP_001263409.1
NM_004793.3 2662 Missense Mutation CAG,GAG Q941E NP_004784.2
XM_011528441.2 2662 Intron XP_011526743.1
Gene
RPL36
Gene Name
ribosomal protein L36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015414.3 2662 Intron NP_056229.2
NM_033643.2 2662 Intron NP_378669.1

View Full Product Details