Product Details
- SNP ID
-
rs146438852
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:8066049 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCCGGTAGGTTCCAGGCCCCGGC[C/T]GCCTCCGCCCCAGCTGCAGGGAGCT
- Phenotype
-
MIM: 602565
MIM: 608529
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCL25
PubMed Links
Gene Details
- Gene
- CCL25
- Gene Name
- C-C motif chemokine ligand 25
There are no transcripts associated with this gene.
- Gene
- FBN3
- Gene Name
- fibrillin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001321431.1 |
8440 |
Missense Mutation |
CAG,CGG |
Q2767R |
NP_001308360.1 |
NM_032447.4 |
8440 |
Missense Mutation |
CAG,CGG |
Q2767R |
NP_115823.3 |
XM_017027372.1 |
8440 |
Missense Mutation |
CAG,CGG |
Q2767R |
XP_016882861.1 |
XM_017027373.1 |
8440 |
Missense Mutation |
CAG,CGG |
Q2747R |
XP_016882862.1 |
XM_017027374.1 |
8440 |
Missense Mutation |
CAG,CGG |
Q2735R |
XP_016882863.1 |
XM_017027375.1 |
8440 |
Missense Mutation |
CAG,CGG |
Q2726R |
XP_016882864.1 |
XM_017027376.1 |
8440 |
Missense Mutation |
CAG,CGG |
Q2724R |
XP_016882865.1 |
XM_017027377.1 |
8440 |
Missense Mutation |
CAG,CGG |
Q1906R |
XP_016882866.1 |
XM_017027378.1 |
8440 |
Missense Mutation |
CAG,CGG |
Q1553R |
XP_016882867.1 |
XM_017027379.1 |
8440 |
Missense Mutation |
CAG,CGG |
Q2809R |
XP_016882868.1 |
View Full Product Details