Product Details
- SNP ID
-
rs146474834
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:53409121 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATTTATACTGGAGAGAAACTACA[C/T]GTGTAATGAGTGTGGTAAGACCTTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF765
PubMed Links
Gene Details
- Gene
- ZNF765
- Gene Name
- zinc finger protein 765
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001040185.1 |
1689 |
Silent Mutation |
CAC,CAT |
H522H |
NP_001035275.1 |
View Full Product Details