Product Details

SNP ID
rs146522353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1384251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCCGAGACCAGGGCACGGCCCG[C/T]GAGGCTGCTCTTGAGATGCCCTGGA
Phenotype
MIM: 601825
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MUM1 PubMed Links

Gene Details

Gene
MUM1
Gene Name
melanoma associated antigen (mutated) 1
There are no transcripts associated with this gene.

Gene
NDUFS7
Gene Name
NADH:ubiquinone oxidoreductase core subunit S7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024407.4 Intron NP_077718.3
XM_005259556.4 Intron XP_005259613.2
XM_017026768.1 Intron XP_016882257.1

View Full Product Details