Product Details

SNP ID
rs146649226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36003344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCATTTATTACACACATCAGACT[A/G]GGGGAAGACCATTGACATAGCTGAG
Phenotype
MIM: 613304 MIM: 612848 MIM: 615535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALKBH6 PubMed Links

Gene Details

Gene
ALKBH6
Gene Name
alkB homolog 6
There are no transcripts associated with this gene.

Gene
SDHAF1
Gene Name
succinate dehydrogenase complex assembly factor 1
There are no transcripts associated with this gene.

Gene
SYNE4
Gene Name
spectrin repeat containing nuclear envelope family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039876.2 981 Missense Mutation CCA,CTA P403L NP_001034965.1
NM_001297735.2 981 Missense Mutation CCA,CTA P290L NP_001284664.1

View Full Product Details