Product Details

SNP ID
rs146680240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45771366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCTTACATGGGAAGGTGGATCC[A/G]TGGCCCGGGGACTGGGGACCCCCGT
Phenotype
MIM: 605377 MIM: 600963
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BHMG1 PubMed Links

Gene Details

Gene
BHMG1
Gene Name
basic helix-loop-helix and HMG-box containing 1
There are no transcripts associated with this gene.

Gene
DMPK
Gene Name
dystrophia myotonica protein kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081560.2 2205 Missense Mutation ACG,ATG T539M NP_001075029.1
NM_001081562.2 2205 Missense Mutation ACG,ATG T539M NP_001075031.1
NM_001081563.2 2205 Missense Mutation ACG,ATG T554M NP_001075032.1
NM_001288764.1 2205 Missense Mutation ACG,ATG T570M NP_001275693.1
NM_001288765.1 2205 Missense Mutation ACG,ATG T455M NP_001275694.1
NM_001288766.1 2205 Intron NP_001275695.1
NM_004409.4 2205 Missense Mutation ACG,ATG T544M NP_004400.4
Gene
SIX5
Gene Name
SIX homeobox 5
There are no transcripts associated with this gene.

View Full Product Details