Product Details

SNP ID
rs146790551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34759082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGTGAGTGAACGATGAACTGTG[A/G]CTGAAGGTCTTTCCACATTCATTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF599 PubMed Links

Gene Details

Gene
ZNF599
Gene Name
zinc finger protein 599
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007248.2 2107 Silent Mutation AGC,AGT S573S NP_001007249.1
XM_017026369.1 2107 Silent Mutation AGC,AGT S536S XP_016881858.1

View Full Product Details