Product Details
- SNP ID
-
rs146801746
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57415909 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAACTCTGAGACTATTTTGGCAGA[A/G]GATGGGATCATCTAGAAGCATTGCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF17
PubMed Links
Gene Details
- Gene
- ZNF17
- Gene Name
- zinc finger protein 17
View Full Product Details