Product Details

SNP ID
rs146876269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52950982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATGTATTGCTTCTGATTAAAGA[C/T]CTTGCCACATACATCACATTTATAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF321P PubMed Links

Gene Details

Gene
ZNF321P
Gene Name
zinc finger protein 321, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF816
Gene Name
zinc finger protein 816
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031665.2 Intron NP_001026835.1
NM_001202456.1 Intron NP_001189385.1
NM_001202457.1 Intron NP_001189386.1
Gene
ZNF816-ZNF321P
Gene Name
ZNF816-ZNF321P readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202473.1 Intron NP_001189402.1

View Full Product Details