Product Details

SNP ID
rs146900491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46040216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCTGGCTTCATGCCTGGGACCT[C/T]GAACCTCACAACTGTCTGGTTCTGA
Phenotype
MIM: 610547
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGFL4 PubMed Links

Gene Details

Gene
IGFL4
Gene Name
IGF like family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002923.1 325 Missense Mutation AAG,GAG K91E NP_001002923.1

View Full Product Details