Product Details

SNP ID
rs146970110
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:603763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGACCCCGAGAAGATCAAGAA[A/G]AAGTATCTGCGCACGTGGTTCGTGG
Phenotype
MIM: 602781
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HCN2 PubMed Links
Additional Information
For this assay, SNP(s) [rs55659726] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HCN2
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001194.3 Intron NP_001185.3

View Full Product Details