Product Details

SNP ID
rs147006672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9126437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAGAAAAAGTGGGGAATTTCCA[C/G]GTCTATGCAGAAGGTCAGCTGTAGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR7G3 PubMed Links

Gene Details

Gene
OR7G3
Gene Name
olfactory receptor family 7 subfamily G member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001958.1 514 Missense Mutation CTG,GTG L172V NP_001001958.1

View Full Product Details