Product Details

SNP ID
rs147046922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46040320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCGCAGAGCCGGGTCTGGTTC[A/C]AGTCTAGGATGACACCGTCATCACA
Phenotype
MIM: 610547
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGFL4 PubMed Links

Gene Details

Gene
IGFL4
Gene Name
IGF like family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002923.1 221 Missense Mutation TGG,TTG W56L NP_001002923.1

View Full Product Details