Product Details

SNP ID
rs147069793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41796726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGAGAATGCATCCCCTGGCAG[A/G]GGCTTCTGCTCACAGGTGAGTGGAG
Phenotype
MIM: 609142
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEACAM3 PubMed Links

Gene Details

Gene
CEACAM3
Gene Name
carcinoembryonic antigen related cell adhesion molecule 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277163.2 137 Missense Mutation AGG,GGG R17G NP_001264092.1
NM_001815.4 137 Missense Mutation AGG,GGG R17G NP_001806.2

View Full Product Details