Product Details

SNP ID
rs147117678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48813247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACCCGTACCCCAGCTCTGCACC[C/G]CCCGTCACGAGCAGTTCAATGCCCG
Phenotype
MIM: 113530 MIM: 612832
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BCAT2 PubMed Links

Gene Details

Gene
BCAT2
Gene Name
branched chain amino acid transaminase 2
There are no transcripts associated with this gene.

Gene
HSD17B14
Gene Name
hydroxysteroid 17-beta dehydrogenase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016246.2 1007 Silent Mutation GGC,GGG G247G NP_057330.2
XM_005258969.3 1007 UTR 3 XP_005259026.1
XM_005258970.3 1007 UTR 3 XP_005259027.1

View Full Product Details