Product Details

SNP ID
rs147175848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38942647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGACCCTCACACTGGAGTGGGT[C/G]ACAAGGGCGCCATAGTGCCCCACCC
Phenotype
MIM: 609094 MIM: 603021
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXO17 PubMed Links

Gene Details

Gene
FBXO17
Gene Name
F-box protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024907.6 930 Silent Mutation GTC,GTG V266V NP_079183.4
NM_148169.2 930 Silent Mutation GTC,GTG V275V NP_680474.1
Gene
MRPS12
Gene Name
mitochondrial ribosomal protein S12
There are no transcripts associated with this gene.

View Full Product Details