Product Details

SNP ID
rs147232734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1440360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTCTCCCGCAGCCCGAGATGAT[C/T]GGCCACTACCTGGGCGAGTTCTCCA
Phenotype
MIM: 612034 MIM: 607430 MIM: 180535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APC2 PubMed Links

Gene Details

Gene
APC2
Gene Name
APC2, WNT signaling pathway regulator
There are no transcripts associated with this gene.

Gene
DAZAP1
Gene Name
DAZ associated protein 1
There are no transcripts associated with this gene.

Gene
RPS15
Gene Name
ribosomal protein S15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018.4 Intron NP_001009.1
NM_001308226.1 Intron NP_001295155.1

View Full Product Details