Product Details
- SNP ID
-
rs147343602
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:18342884 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATTTGGCCCTTTTGGGGAACACAC[C/T]GTGAACGCCAGTTGGATTGCAGTTC
- Phenotype
-
MIM: 610694
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PGPEP1
PubMed Links
Gene Details
- Gene
- PGPEP1
- Gene Name
- pyroglutamyl-peptidase I
There are no transcripts associated with this gene.
View Full Product Details