Product Details

SNP ID
rs147396237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15053503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGAGAAATATGATATGTCAGGT[G/T]CCCGCCTGGCCCTAATACTGTGTGT
Phenotype
MIM: 605848
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CASP14 PubMed Links

Gene Details

Gene
CASP14
Gene Name
caspase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012114.2 261 Missense Mutation GCC,TCC A17S NP_036246.1
XM_011527861.1 261 Missense Mutation GCC,TCC A17S XP_011526163.1

View Full Product Details