Product Details
- SNP ID
-
rs147428178
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:2389854 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAGGAAGTCCCCGCTCTGGATGC[C/T]GCGTGCTGTCGAGCGGCCAGCATTG
- Phenotype
-
MIM: 610477
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMPRSS9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8100380,rs891174] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMPRSS9
- Gene Name
- transmembrane protease, serine 9
View Full Product Details