Product Details

SNP ID
rs147432413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57547064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACACTTGTAGGGCATCTCCCCAG[G/T]GTGGATGACAGAGTGCTGAATGAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF549 PubMed Links

Gene Details

Gene
ZNF549
Gene Name
zinc finger protein 549
There are no transcripts associated with this gene.

Gene
ZNF550
Gene Name
zinc finger protein 550
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277090.1 1358 Missense Mutation ACT,CCT T394P NP_001264019.1
NM_001277091.1 1358 Missense Mutation ACT,CCT T394P NP_001264020.1
NM_001277092.1 1358 Missense Mutation ACT,CCT T394P NP_001264021.1
NM_001277093.1 1358 Intron NP_001264022.1
XM_011526567.2 1358 Missense Mutation ACT,CCT T394P XP_011524869.1
XM_011526568.2 1358 Missense Mutation ACT,CCT T385P XP_011524870.1
XM_017026401.1 1358 Missense Mutation ACT,CCT T394P XP_016881890.1

View Full Product Details