Product Details

SNP ID
rs147501741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57129013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAATCTCAGCAACCCATGAAAT[A/C]TGATGATGATTGGAGTGTGTTTGAA
Phenotype
MIM: 609546
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
USP29 PubMed Links

Gene Details

Gene
USP29
Gene Name
ubiquitin specific peptidase 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020903.2 694 Missense Mutation TAT,TCT Y113S NP_065954.1
Gene
ZIM3
Gene Name
zinc finger imprinted 3
There are no transcripts associated with this gene.

View Full Product Details