Product Details

SNP ID
rs147548953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15013751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAGAGGAAAATGGAGAGAGATA[C/T]GGAAAAATCAGAGGTCCTACTCAAG
Phenotype
MIM: 600637
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC105 PubMed Links

Gene Details

Gene
CCDC105
Gene Name
coiled-coil domain containing 105
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173482.2 749 Missense Mutation ACG,ATG T217M NP_775753.2
Gene
SLC1A6
Gene Name
solute carrier family 1 member 6
There are no transcripts associated with this gene.

View Full Product Details