Product Details

SNP ID
rs147598079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32598992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTTCTGAGTGATTTAAAAATGC[A/G]GTAGGGTCCGGGCACGGTGGCTCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD27 PubMed Links

Gene Details

Gene
ANKRD27
Gene Name
ankyrin repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032139.2 Intron NP_115515.2
XM_006723412.2 Intron XP_006723475.1
XM_017027349.1 Intron XP_016882838.1

View Full Product Details