Product Details

SNP ID
rs147599228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14121567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGCTTCATGGGCGGGTTCTCAC[A/G]CAGCTCAGGGTTGAGGTACTCGTTG
Phenotype
MIM: 609190 MIM: 601639
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASF1B PubMed Links

Gene Details

Gene
ASF1B
Gene Name
anti-silencing function 1B histone chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018154.2 539 Missense Mutation CGT,TGT R123C NP_060624.1
Gene
PRKACA
Gene Name
protein kinase cAMP-activated catalytic subunit alpha
There are no transcripts associated with this gene.

View Full Product Details