Product Details

SNP ID
rs147630109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46411542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTGCCCTCTGATTATCTGCACC[C/T]TGGACAGCTGGGGCCCTTTCCCTCT
Phenotype
MIM: 614145
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC8 PubMed Links

Gene Details

Gene
CCDC8
Gene Name
coiled-coil domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032040.4 2121 Silent Mutation CAA,CAG Q423Q NP_114429.2

View Full Product Details