Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001321411.1 | 1238 | Silent Mutation | AGC,AGT | S339S | NP_001308340.1 |
NM_031917.2 | 1238 | Silent Mutation | AGC,AGT | S339S | NP_114123.2 |
XM_005260091.3 | 1238 | Silent Mutation | AGC,AGT | S420S | XP_005260148.1 |
XM_011528347.2 | 1238 | Silent Mutation | AGC,AGT | S339S | XP_011526649.1 |
XM_011528348.2 | 1238 | Silent Mutation | AGC,AGT | S406S | XP_011526650.1 |
XM_011528349.2 | 1238 | Silent Mutation | AGC,AGT | S406S | XP_011526651.1 |
XM_011528350.2 | 1238 | Silent Mutation | AGC,AGT | S339S | XP_011526652.1 |
XM_017027347.1 | 1238 | Silent Mutation | AGC,AGT | S339S | XP_016882836.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001308277.1 | 1238 | Intron | NP_001295206.1 | ||
NM_018381.3 | 1238 | Intron | NP_060851.2 | ||
XM_011528121.1 | 1238 | Intron | XP_011526423.1 | ||
XM_011528122.1 | 1238 | Intron | XP_011526424.1 | ||
XM_017026934.1 | 1238 | Intron | XP_016882423.1 |