Product Details

SNP ID
rs147710123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1388848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGCCAGCACCCAGCCTGCCCT[A/G]CCAAAGGCCAGAGCCGTGGCTCCCA
Phenotype
MIM: 601240 MIM: 601825
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAMT PubMed Links

Gene Details

Gene
GAMT
Gene Name
guanidinoacetate N-methyltransferase
There are no transcripts associated with this gene.

Gene
NDUFS7
Gene Name
NADH:ubiquinone oxidoreductase core subunit S7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024407.4 181 Silent Mutation CTA,CTG L46L NP_077718.3
XM_005259556.4 181 Silent Mutation CTA,CTG L46L XP_005259613.2
XM_017026768.1 181 Silent Mutation CTA,CTG L131L XP_016882257.1

View Full Product Details