Product Details

SNP ID
rs147713850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10134265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAACATACAAAGCTTGATCTCCA[A/G]GCCAATGGCTTTGGCCAGGGGCGGT
Phenotype
MIM: 126375
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNMT1 PubMed Links

Gene Details

Gene
DNMT1
Gene Name
DNA (cytosine-5-)-methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130823.2 5052 Silent Mutation CTG,TTG L1606L NP_001124295.1
NM_001318730.1 5052 Silent Mutation CTG,TTG L1593L NP_001305659.1
NM_001318731.1 5052 Silent Mutation CTG,TTG L1485L NP_001305660.1
NM_001379.3 5052 Silent Mutation CTG,TTG L1590L NP_001370.1

View Full Product Details