Product Details

SNP ID
rs147737549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40380467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGTCATCCTCAGGCGAGACCTG[C/G]CTCAGCCGAATGAGGTTGCTGAAGT
Phenotype
MIM: 611712 MIM: 615698
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HIPK4 PubMed Links

Gene Details

Gene
HIPK4
Gene Name
homeodomain interacting protein kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144685.4 1818 Missense Mutation AGC,AGG S508R NP_653286.2
XM_006723036.2 1818 Missense Mutation AGC,AGG S508R XP_006723099.1
Gene
PLD3
Gene Name
phospholipase D family member 3
There are no transcripts associated with this gene.

View Full Product Details