Product Details

SNP ID
rs148112821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13963573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACTTGAGGAGGAAGAGCTTGGC[G/T]GCCTTGGGGAAGCCGGCGCTGCCCT
Phenotype
MIM: 600006
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DCAF15 PubMed Links

Gene Details

Gene
DCAF15
Gene Name
DDB1 and CUL4 associated factor 15
There are no transcripts associated with this gene.

Gene
RFX1
Gene Name
regulatory factor X1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002918.4 3311 Silent Mutation GCA,GCC A845A NP_002909.4
XM_011528165.2 3311 Silent Mutation GCA,GCC A955A XP_011526467.1
XM_011528166.2 3311 Silent Mutation GCA,GCC A955A XP_011526468.1
XM_011528167.2 3311 Silent Mutation GCA,GCC A955A XP_011526469.1
XM_011528168.2 3311 Silent Mutation GCA,GCC A857A XP_011526470.1
XM_011528169.2 3311 Silent Mutation GCA,GCC A846A XP_011526471.1
XM_011528170.2 3311 Silent Mutation GCA,GCC A846A XP_011526472.1

View Full Product Details