Product Details

SNP ID
rs148114033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:687015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCAGGAGCTGTTGCAGACGTC[C/T]GGGTCCAGCACTCGGACCTTGGCCT
Phenotype
MIM: 605343
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FSTL3 PubMed Links

Gene Details

Gene
FSTL3
Gene Name
follistatin like 3
There are no transcripts associated with this gene.

Gene
PRSS57
Gene Name
protease, serine 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308209.1 583 Silent Mutation CCA,CCG P184P NP_001295138.1
NM_214710.4 583 Silent Mutation CCA,CCG P185P NP_999875.1

View Full Product Details