Product Details

SNP ID
rs148131005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49347180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGAGCACTTTTGATTTTGCTGT[A/G]AGAACTCACCCAGAACTTGACCAGG
Phenotype
MIM: 151523 MIM: 601729
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD37 PubMed Links

Gene Details

Gene
CD37
Gene Name
CD37 molecule
There are no transcripts associated with this gene.

Gene
TEAD2
Gene Name
TEA domain transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256658.1 Intron NP_001243587.1
NM_001256659.1 Intron NP_001243588.1
NM_001256660.1 Intron NP_001243589.1
NM_001256661.1 Intron NP_001243590.1
NM_001256662.1 Intron NP_001243591.1
NM_003598.1 Intron NP_003589.1
XM_005259334.3 Intron XP_005259391.1
XM_006723424.1 Intron XP_006723487.1
XM_006723428.2 Intron XP_006723491.1
XM_006723429.2 Intron XP_006723492.1
XM_011527398.1 Intron XP_011525700.1
XM_011527399.2 Intron XP_011525701.1
XM_011527400.2 Intron XP_011525702.1
XM_011527401.1 Intron XP_011525703.1
XM_011527402.2 Intron XP_011525704.1
XM_011527403.1 Intron XP_011525705.1
XM_011527404.1 Intron XP_011525706.1
XM_011527405.2 Intron XP_011525707.1
XM_011527406.2 Intron XP_011525708.1

View Full Product Details